missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: m LES Endo Agar

For enumerating coliforms in water by membrane filtration

€ 112.20 - € 297.00

Specifications

Certifications/Compliance CE, COMPF, SMD, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type mLES Endo Agar
Product Type Agar
View More Specs

Products 2
Product Code Brand For Use With (Application) Quantity Price Quantity & Availability  
Product Code Brand For Use With (Application) Quantity Price Quantity & Availability  
11743483
View Documents Promotion Details
BD
273620
For enumerating Coliforms in Water by membrane filtration 500 g
€ 297.00
500g
View alternative products
Save Up to 
»
Estimated Shipment: 15-04-2024
to see stock.
Add to basket
 
11723483
View Documents Promotion Details
BD
273610
Enumerating Coliforms in Water by membrane filtration 100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Formulation: Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g

Certifications

CE, COMPF, SMD, SMWW

Specifications

Specifications

CE, COMPF, SMD, SMWW
Dehydrated
Agar
Bottle
<30°C
mLES Endo Agar
Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g/L, Basic Fuchsin 0.8g/L, Agar 15g/L
Videos
SDS
Documents

Documents

Product Certifications