Learn More
BD Difco™ Dehydrated Culture Media: m Staphylococcus Broth
Click to view available options
:
M-Staphylococcus Broth; 500g
Specifications
Specifications
| Certifications/Compliance | SMWW |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | M-Staphylococcus Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Caseing/L, Yeast extract 2.5g/L, Lactose 2g/L, Mannitol 10g/L, Dipotassium Phosphate 5g/L, Sodium Chloride 75g/L |
| For Use With (Application) | For isolating Staphylococci by the membrane filtration technique |
| Packaging Type | Bottle |
| Quantity | 500 g |