Learn More
BD Difco™ Dehydrated Culture Media: Mueller Hinton Agar
For antimicrobial susceptibility testing of microorganisms by agar diffusion methods.  BD Difco™ Dehydrated Culture Media: Mueller Hinton Agar is part of a large selection of dehydrated media.
Brand:  BD 225250
Product Code. 11762804
  • € 150.00 / 500g
Edge
Description
Description
  • Dehydrated
  • Culture Media
  • Mueller Hinton Agar