Learn More
BD Difco™ Dehydrated Culture Media: Pantothenate Medium
Click to view available options
:
Pantothenate Medium AOAC USP; 100g
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Certifications
AOAC, USP
Specifications
Specifications
| Certifications/Compliance | AOAC, USP |
| Content And Storage | 2°C to 30°C |
| Form | Dehydrated |
| Media Type | Pantothenate Medium AOAC USP |
| Product Type | Pantothenate Assay Medium |
| Formulation | Dextrose 40g/L, Sodium Acetate 20g/L, Vitamin assay Casamino Acids 10g/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, L-Cystine 0.4g/L, L-Tryptophan 0.1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese |
| For Use With (Application) | Determination of concentration of pantothenic Acid and Pantothenate by the Ficrobiological assay technique |
| Packaging Type | Bottle |
| Quantity | 100 g |