missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Tryptose Blood Agar Base
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
Description
Used with blood for isolation and cultivation of fastidious microorganisms as well as determination of hemolytic reactions
Certifications
BAM
Specifications
Specifications
| Certifications/Compliance | BAM |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Tryptose Blood Agar Base |
| Product Type | Agar |
| Formulation | Tryptose 10g/L, Beef extract 3g/L, Sodium Chloride 5g/L, Agar 15g/L |
| For Use With (Application) | For used with Blood in isolating cultivating and determining the hemolytic reactions |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction