Learn More
BD Difco™ HYcheck™ Contact Slides: Yeasts and Molds with TTC
To assess microbiological contamination of surfaces or fluids. BD Difco™ HYcheck™ Contact Slides: Yeasts and Molds with TTC are designed for environmental testing.
Brand: BD 290007
Additional Details : Weight : 0.80000kg
Description
- Designed for environmental testing
- Double sided with Tryptic Soy Agar with 0.01% TTC and Rose Bengal Chloramphenicol Agar
- Hinged paddle bends for easy sampling
Order Info
Ten units per pack.
Specifications
2°C to 8°C | |
United States | |
To assess microbiological contamination of surfaces or fluids | |
10 U |
Solid | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L, 0.01% TTC, Papaic digest of Soybean Meal 5g/L, Dextrose 10g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Rose Bengal 0.05g/L, Agar 15g/L, Chloramphenicol 0.05g/L | |
Slide |