missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Rat Anti-Human CXCR5 (CD185)

Product Code. 15874779
Change view
Click to view available options
Sufficient For:
100 Tests
25 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
15874779 100 Tests Each
15844829 25 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15874779 Supplier BD Supplier No. 566142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In peripheral blood, CXCR5 expression is restricted to B lymphocytes and a small subset of CD4+ and CD8+ lymphocytes. BD Horizon™ BV480 Rat Anti-Human CXCR5 (CD185) has restricted expression pattern of CXCR5 on B cells and follicular T helper cells (Tfh) that suggests this receptor functions as a regulator of B and T cell migration.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Rat Anti-Human CXCR5 (CD185)
Quantity 5 μL
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Sufficient For 100 Tests
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.