missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Rat Anti-Mouse CD45

Product Code. 15834809
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15834809

Brand: BD 566168

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BD Horizon - BV480 Rat Anti-Mouse CD45 - CD45 is a transmembrane glycoprotein which is expressed at high levels on cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Rat Anti-Mouse CD45
Quantity 25 μg
For Use With (Application) Flow Cytometry
Concentration 0.2 mg/mL
Cross Reactivity QC Testing: Mouse
Form BV480
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.