missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human IL-12 (p40) ELISA Kit II
Description
Includes
Antibody Coated Wells, Detection Antibody, Standards, Enzyme Concentrate (250x), Standard/Sample Diluent, ELISA Diluent, Wash Concentrate (20x), TMB One-Step Substrate Reagent, Stop Solution, Plate Sealers
- Potent regulator of cell-mediated immune responses
- Secreted by activated B lymphocytes and macrophages
Applications:
ELISA
Specifications
Specifications
| Antigen | IL-12 (p40) |
| Cross Reactivity | Human |
| Includes | Wash concentrate (20X), stop solution, Enzyme concentrate Streptavidin HRP, Human IL-12 (p40) Antibody coated plate, detection Antibody Biotin Anti-Human IL-12 (p40), Human IL-12 (p40) standard, ELISA Diluent, standard/Sample Diluent, TMB one-step Substrate reagent |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Quantitative determination of Human IL-12 (p40), ELISA |
| Product Type | Human IL-12 (p40) ELISA kit II |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?