Learn More
BD Human TNF ELISA Set
An immunoassay with different sensitivity and dynamic range
Brand: BD 555212
Additional Details : Weight : 0.16000kg
Includes: Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
Description
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human TNF in serum, plasma, and cell culture supernatants
Enzyme-linked immunosorbent assay
Specifications
7.8 to 500pg/mL | |
20 96-well Plates | |
anti-Human TNF monoclonal (capture) Antibody, Biotinylated anti-Human TNF monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human TNF (standard), Streptavidin-HRP Conjugate |
Human TNF ELISA set | |
To develop ELISA for natural or recombinant human TNF in serum, plasma and cell culture supernatants | |
2°C to 8°C |
Research Use Only