Learn More
BD Luer-Lok™ Access Device
Product Code. 10782337
Change view
Click to view available options
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
10782337 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10782337 Supplier BD Supplier No. 36490200
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Use for needleless collection of blood. BD Vacutainer™ Luer-Lok™ Access Device is a direct access collection device.
  • For needleless collection of blood
  • Holder with preattached multiple sample, male adapter
  • Compatible with female luer connections or IV ports designed for luer access
  • Allows specimens to be collected into an evacuated tube
  • Has blue color-coded threads to provide easy differentiation from other holder-based products
  • Provides the security of a threaded locking luer connection, replacing luer slip devices
  • Compatible with luer-activated ports for closed, direct urine sampling found in some Foley catheters (such as the Bard™ E-Z™ Lock Sampling Port)
  • Sterile; latex-free
TRUSTED_SUSTAINABILITY
Specifications
Connectors Male Luer
For Use With (Application) For needle-less collection of Blood
For Use With (Equipment) Luer activated ports for closed, direct Urine sampling found in some Foley catheters
Latex Free Yes
Product Type Blood Collection Access Device
Sterility Sterile
Non-returnable medical device