Learn More
BD Luer-Lok™ Access Device
Use for needleless collection of blood. BD Vacutainer™ Luer-Lok™ Access Device is a direct access collection device.
Brand: BD 36490200
Additional Details : Weight : 1.50000kg
Description
- For needleless collection of blood
- Holder with preattached multiple sample, male adapter
- Compatible with female luer connections or IV ports designed for luer access
- Allows specimens to be collected into an evacuated tube
- Has blue color-coded threads to provide easy differentiation from other holder-based products
- Provides the security of a threaded locking luer connection, replacing luer slip devices
- Compatible with luer-activated ports for closed, direct urine sampling found in some Foley catheters (such as the Bard™ E-Z™ Lock Sampling Port)
- Sterile; latex-free
Specifications
Male Luer | |
Blood Collection Access Device | |
For needle-less collection of Blood |
Sterile | |
Yes | |
Luer activated ports for closed, direct Urine sampling found in some Foley catheters |
Non-returnable medical device