Learn More
BD Mouse IFN-γ ELISA Kit II
For quantitative determination of mouse IFN-γ in serum, plasma and cell culture supernatant
Brand: BD 558258
Additional Details : Weight : 0.61200kg
Includes: Stop solution, Standard/sample diluent, TMB one-Step substrate reagent, detection antibody biotin anti-mouse IFN-gamma, mouse IFN-gamma standard, ELSIA diluent III, Enzyme concentrate streptavidin-HRP, wash concentrate, mouse IFN-g antibody coated plate
Description
The BD OptEIA™ test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay).
- 30 kD homodimeric protein, as shown by SDS-PAGE analysis
- Utilizes a monoclonal antibody specific for mouse IFN-γ coated on a 96-well plate
Applications:
Enzyme-linked immunosorbent assay
Specifications
IFN-gamma | |
Stop solution, standard/Sample diluent, TMB one-step Substrate reagent, detection Antibody Biotin Anti-Mouse IFN-gamma, Mouse IFN-gamma standard, ELISA diluent III, Enzyme concentrate Streptavidin HRP, wash concentrate (20X), Mouse IFN-g Antibody coated plate | |
Serum, Plasma, Cell Culture Supernatant | |
Quantitative determination of Mouse IFN-gamma, ELISA |
Mouse | |
2 Plates | |
2°C to 8°C | |
Mouse IFN-gamma ELISA kit II |
Research Use Only