Learn More
BD Prepared Bottled Media: Tryptic Soy Broth, Wide Mouth Bottle
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Ready for immediate use
- Sterility Assurance Level of 10-6 reduces risk of false positives
- Terminally autoclaved
- Conforms to Harmonized USP, EP, and JP requirements for sterility tests and to CLSI requirements for recovery
Specifications
Specifications
| Cap Type | Lug cap |
| Color | Amber |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Tryptic Soy Broth |
| Fill Volume | 100 mL |
| Packaging Type | Bottle |
| Quantity | 100 mL |
| Height (Metric) | 130 mm |
| Diameter (Metric) | 48 mm |