missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Mouse Interleukin-5 (IL-5)

Product Code. 15877788
Click to view available options
Quantity:
5 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15877788

Brand: BD 554581

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lymphokine for activities related to eosinophil differentiating factor, B cell growth factor II, T cell-replacing factor

  • 35-37kD homodimer
  • Induces eosinophil differentiation
  • Promotes survival and activation of eosinophil
  • Stimulates generation of cytotoxic T cell
  • Supports growth and differentiation of pre-activated B cells and hematopoietic cells
  • Reactivity: mouse
  • Storage buffer: frozen aqueous buffered solution containing BSA and glycerol

ELISA standard, functional assay, blocking

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100μg/mL
For Use With (Application) Blocking Agent, ELISA Standard, Functional Assay
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Molecular Weight (g/mol) 35 to 37
Name Mouse IL-5 Recombinant Protein
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Solution

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.