Learn More
BD ROCK Inhibitor (Y-27632)
Click to view available options
Quantity:
1 mg
Unit Size:
Each
Description
The ROCK inhibitor, Y-27632, is a selective inhibitor of the p160-Rho-associated coiled kinase (ROCK) and is a factor that enhances embryonic stem cell survival upon single cell dissociation. In addition to increasing cell recovery after dissociation, Y-27632 has been used for various other applications in stem cell research including cryopreservation, sorting, reprogramming, transplantation, and differentiation.
Cell Culture
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | ROCK inhibitor (Y-27632) |
| Quantity | 1 mg |