Learn More
BD Streptavidin Particles Plus - DM
Product Code. 15879058
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15879058 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15879058 Supplier BD Supplier No. 557812
For positive selection or depletion of leukocyte subpopulations using the BD IMagnet™
The small size of BD IMag particles allows the positive fraction to be further evaluated in downstream applications.
  • Magnetic nanoparticles
  • Have streptavidin covalently conjugated to their surface
  • Allow positive fraction to be further evaluated in downstream applications such as flow cytometry
  • Storage Buffer: Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Cell separation
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mL
Content And Storage 4°C
For Use With (Application) Cell separation, for positive selection or depletion of leukocyte subpopulations using the BD IMagnet