missing translation for 'onlineSavingsMsg'
Learn More

BD1063 dhydrochloride, MedChemExpress

Product Code. 16483487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483487 50 mg 50mg
16449947 5 mg 5mg
16437468 10 mg 10mg
16423596 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483487 Supplier Medchem Express Supplier No. HY18101A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BD1063 dhydrochloride is a potent and selective sigma 1 receptor antagonist.

BD1063 dhydrochloride is a potent and selective sigma 1 receptor antagonist.

Specifications

Chemical Name or Material BD1063 dhydrochloride
Physical Form Solid
CAS 206996-13-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H20Cl4N2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information H2O : 100 mg/mL (288.92 mM; Need ultrasonic)
SMILES CN1CCN(CCC2=CC=C(Cl)C(Cl)=C2)CC1.Cl.Cl
Molecular Weight (g/mol) 346.12
Formula Weight 346.12
Percent Purity 96.77%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.