missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDH2 Antibody (OTI2G1), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72067MFV500
This item is not returnable.
View return policy
Description
BDH2 Monoclonal antibody specifically detects BDH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| BDH2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI2G1 | |
| Flow Cytometry | |
| 3-hydroxybutyrate dehydrogenase, type 2, dehydrogenase/reductase (SDR family) member 6, Dehydrogenase/reductase SDR family member 6, DHRS6, EC 1.1.1.-, EC 1.1.1.30, EFA6R, FLJ13261, Oxidoreductase UCPA, PRO20933, R-beta-hydroxybutyrate dehydrogenase, SDR15C1,3-hydroxybutyrate dehydrogenase type 2, short chain dehydrogenase/reductase family 15C, member 1, UCPA-OR, UNQ6308 | |
| Full length human recombinant protein of human BDH2(NP_064524) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 56898 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction