missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDNF Antibody (35928.11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10505H
This item is not returnable.
View return policy
Description
BDNF Monoclonal antibody specifically detects BDNF in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| BDNF | |
| Monoclonal | |
| HRP | |
| abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
| Recombinant human BDNF (rhBDNF) expressed in Sf 21 insect cells. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Neuroscience | |
| 627 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 35928.11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction