missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BDP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562169
This item is not returnable.
View return policy
Description
Immunogen sequence: GSSSGTNTSS PRISPATTLR KGDGAPVTTV PVPNRLAVPV TLASQQAGTR TAALEQLR Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 78%.
This gene encodes a member of the ARID family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.
Specifications
| BDP | |
| Polyclonal | |
| Unconjugated | |
| ARID3B | |
| ARID domain-containing protein 3B; ARID3B; AT rich interactive domain 3B (Bright like); AT rich interactive domain 3B (BRIGHT- like); AT rich interactive domain 3B (BRIGHT-like); AT-rich interaction domain 3B; AT-rich interactive domain-containing protein 3B; Bdp; bright and dead ringer gene product homologous protein Bdp; bright and dead ringer protein; Bright-like; Bright-like protein; dead ringer homolog 2; Dri2; DRIL2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVW6 | |
| ARID3B | |
| Recombinant protein corresponding to Human BDP. Recombinant protein control fragment (Product #RP-99932). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction