missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Beaded Rim Culture Media Bottle
Merk: Duran™ 214513907
Overige gegevens : Gewicht : 1.35000kg
Specificatie
300 mL | |
Preparation Of Cultures In Nutrient Media | |
10Pack | |
70mm |
42mm | |
Glass | |
Media Culture Bottle | |
168mm |