Learn More
Pinnacle Brush™ Beaker Brushes
Click to view available options
Length (Metric):
26 cm
27 cm
36.5 cm
39 cm
49 cm
Bristle Material:
Fiber
Nylon
Unit Size:
Pack of 10
Description
- Galvanized steel handle
- Nylon bristles
Specifications
Specifications
| Product Type | Bottle Brush |
| Bristle Material | Nylon |
| Handle Material | Metal |
| Diameter (Metric) | 5.4 cm |
| Length (Metric) | 36.5 cm |
| Length (Metric) Brush | 12.5 cm |
| Length (Metric) Handle | 24 cm |
| Quantity | 10 Pk. |
| Size | Brush Circumference: 17 cm |