Learn More
BUCHI Beaker and Drying Flasks for Rotary Evaporators
Product Code. 15520590 Shop All Buchi Products
Change view
Click to view available options
Type:
Beaker Flask
Drying Flask
Volume (Metric):
500 mL
1500 mL
4000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Volume (Metric) Unit Size
15520590 Drying Flask 1500 mL Each
15510590 Drying Flask 500 mL Each
15590580 Beaker Flask 500 mL Each
17214664 Beaker Flask 4000 mL Each
17224664 Beaker Flask 1500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15520590 Supplier BUCHI Supplier No. 11063160
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Special shaped flasks for large samples and powder drying.
  • For use with the rotary evaporators
  • Available in multiple sizes with different standard joints
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Type Drying Flask
Volume (Metric) 1500 mL