Learn More
Buerkle™ Beakers with Handle and Transparent Graduations
Catalog Number p-8002927
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
5000 mL
Use Buerkle™ Beakers with Handle and Transparent Graduations offer excellent chemical resistance that makes them usable for a wide range of measuring needs in the laboratory. The rectangular handle offers reliable support and dispensing control.
  • PP (polypropylene)
  • Translucent
  • Autoclavable
  • Graduated for measuring
Color Translucent
Diameter (Metric) Base 40 mm
Diameter (Metric)Top 50 mm
Capacity (Metric) 100 mL
Material Polypropylene
For Use With (Application) Suitable for Use with Foodstuffs
Height (Metric) 80 mm
Includes Handle
Type Beaker
Autoclavable Autoclavable
Graduated Yes
Scale Transparent Scale