missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclin 2 Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18252JF635
This item is not returnable.
View return policy
Description
Beclin 2 Polyclonal antibody specifically detects Beclin 2 in Human,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Beclin 2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 635 | |
| BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1 | |
| Synthetic peptide from the C-terminal of Human Beclin 2 (aa. 338-349) | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Tumor Suppressors | |
| 441925 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction