missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Beclin 2 Polyclonal Antibody

Description
Suggested positive control: 293 whole cell lysate, HEK293 kidney cell lysates and MCC827 lung cell lysates. Does not appear to work with A431 lysates.
Involved in 2 distinct lysosomal degradation pathways: acts as a regulator of autophagy and as a regulator of G-protein coupled receptors turnover. Regulates degradation in lysosomes of a variety of G-protein coupled receptors via its interaction with GPRASP1/GASP1. [UniProt].
Specifications
Specifications
| Antigen | Beclin 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | Becn2 |
| Gene Accession No. | A8MW95, P0DM65 |
| Gene Alias | Bcl-2; BCS2; beclin 1, autophagy related, pseudogene 1; beclin 2; beclin-1 autophagy-related pseudogene 1; Beclin-1-like protein 1; Beclin-2; BECN1L1; BECN1P1; BECN2; Gm104; PFM 2; PFM2; PRDM 5; PRDM5; PRDM5 protein; RGD1563293; similar to Beclin 1 (Coiled-coil myosin-like BCL2-interacting protein) |
| Gene Symbols | Becn2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?