missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclometasone EP Impurity C, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Beclometasone EP Impurity C
Specifications
Specifications
| Chemical Name or Material | Beclometasone 21-Butyrate 17-Propionate |
| Analyte or Component Names | Beclometasone 21-Butyrate 17-Propionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H39 Cl O7 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 9-Chloro-11β-hydroxy-16β-methyl-3,20-dioxo-17-(propanoyloxy)pregna-1,4-dien-21-yl Butanoate (Beclometasone 21-Butyrate 17-Propionate), Beclometasone Dipropionate Imp. C (EP), 9-Chloro-11beta-hydroxy-16beta-methyl-3,20-dioxo-17-(propanoyloxy)pregna-1,4-dien-21-yl Butanoate, Beclometasone 21-Butyrate 17-Propionate, Beclometasone Dipropionate Impurity C |
| InChI Formula | InChI=1S/C9H15NO/c11-8-6-9(7-10-8)4-2-1-3-5-9/h1-7H2,(H,10,11) |
| SMILES | CCCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] butanoate |
| Molecular Weight (g/mol) | 535.07 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction