Learn More
BEGAIN Monoclonal Antibody (OTI6E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810836
This item is not returnable.
View return policy
Description
May sustain the structure of the postsynaptic density (PSD).Specifications
| BEGAIN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BUH8 | |
| BEGAIN | |
| Human recombit protein fragment corresponding to amino acids 374-517 of human BEGAIN produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6E7 | |
| Unconjugated | |
| BEGAIN | |
| brain enriched guanylate kinase associated | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57596 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |