Learn More
BEGAIN Monoclonal Antibody (OTI6E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810836S
This item is not returnable.
View return policy
Description
BEGAIN Monoclonal antibody specifically detects BEGAIN in Human samples. It is validated for Western BlotSpecifications
| BEGAIN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BUH8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57596 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6E7 | |
| Unconjugated | |
| BEGAIN | |
| BEGAIN; BM948371; brain enriched guanylate kinase associated; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated homolog; brain-enriched guanylate kinase-associated protein; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated protein; Gm897; KIAA1446 | |
| Human recombinant protein fragment corresponding to amino acids 374-517 of human BEGAIN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |