missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BEGAIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599594
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BEGAIN.
May sustain the structure of the postsynaptic density (PSD). Target protein binds the guanylate kinase domain of PSD-95/SAP90, may play a role in the post-synaptic membrane.
Specifications
| BEGAIN | |
| Polyclonal | |
| Unconjugated | |
| BEGAIN | |
| BEGAIN; BM948371; brain enriched guanylate kinase associated; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated homolog; brain-enriched guanylate kinase-associated protein; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated protein; Gm897; KIAA1446 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57596 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BUH8 | |
| BEGAIN | |
| A synthesized peptide derived from human BEGAIN(Accession Q9BUH8), corresponding to amino acid residues L195-P245. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction