Learn More
Bel-Art™ Bel-Blotter™ 96-Well Replicating Tool
Product Code. 10368204 Shop All SP Scienceware Products
Click to view available options
:
Replicating Tool
Product Code. 10368204
Brand: Bel-Art™ F378760002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to do tedious tasks with a minimum of effort
  • The 96 open-ended pipette tips of this tool fit all types of 96 well plates from flat, vee or round bottom plates to 0.2mL thin walled PCR plates and tubes
  • Can be used for replication of recombinant DNA libraries, inoculation of filters for colony hybridization, PCR, phage typing and other applications
  • Made of polycarbonate plastic, it is easy-to-use, reusable and autoclavable
Specifications
Autoclavable Autoclavable
Material Polycarbonate
Sterility Non-sterile
For Use With (Application) Increase Productivity, Minimize Effort
Quantity 1 Pack
Color Transparent
Volume (Metric) 0.2 mL
Length (Metric) 12.8 cm
Type 96-Well Replicating Tool
Disposable Reusable