Learn More
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
1 mL
5 mL
Unit Size:
Each
Description
- Made from clear acrylic
- Filler hole screws on each half cavity are fluoropolymer-coated stainless steel
- Volume shown is for one half cell (double for total cell volume)
- A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves
Specifications
Specifications
| Product Type | Dialysis Cell |
| Material | Acrylic |
| Capacity (Metric) | 1 mL |
| Color | Transparent |
| Quantity | Each |
| For Use With (Application) | Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket |