Learn More
Bel-Art™ SP Scienceware™ Labels for Protective Labeling Tape System
Labels for use with Bel Art Scienceware Protective Labeling Tape System (cat. no. 1186750)
Brand: Bel-Art™ F13455-0015
Additional Details : Weight : 0.25000kg
Description
- White paper labels accept any kind of marker; perfect for use under the clear tape to label labware, bottles or samples
Specifications
White | |
Paper | |
Use to label labware, bottles or samples | |
3 in. | |
1.5 in. |
Label | |
Label | |
1000/Pk. | |
7.62 cm |