Learn More
Bel-Art™ Labels for Protective Labeling Tape System
Product Code. 10750792 Shop All SP Scienceware Products
Change view
Click to view available options
Width (English):
1 in.
1.5 in.
Length (English):
1.5 in.
3 in.
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Length (English) unitSize
10750792 1.5 in. 1.5 in. Pack of 1000
10443321 1 in. 3 in. Pack of 1000
10392811 1.5 in. 3 in. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10750792 Supplier Bel-Art™ Supplier No. F134550005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Labels for use with Bel-Art™ Protective Labeling Tape System (cat. no. 1186750)
  • White paper labels accept any kind of marker; perfect for use under the clear tape to label labware, bottles or samples
Spécification
Adhesive Type Non-adhesive
Color White
Display Consignment
Finish Matte
Format Label
Material Paper
Printable Sides Double-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Use to label labware, bottles or samples
Quantity 1000/Pk.
Size 1.5 in.
Dimensions (LxW) 38 x 12.5 mm
Length (English) 1.5 in.
Length (Metric) 3.81 cm
Width (English) 1.5 in.
Width (Metric) 12.5 mm