Learn More
Bel Art™ No-Wire™ Microcentrifuge Tube Racks
Product Code. 30306875 Shop All SP Scienceware Products
Change view
Click to view available options
Positions:
42
Holds:
Microtubes (1.5 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Holds unitSize
30306875 42 Microtubes (1.5 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30306875 Supplier Bel Art™ Supplier No. F188380015
Only null left
Add to basket
Add to basket
Open sides optimize use in crushed ice or water baths
  • Molded polypropylene racks sink in water bath and will not tilt
  • Stackable compact size conserves valuable space
  • Autoclavable at 121°C (250°F)
  • Racks are shipped flat: easy-to-assemble
Order Info
Available in two sizes to accommodate most popular tube sizes. Dimensions (L x W x H): 5 x 4 x 1.7 in. (13 x 11 x 4.3cm)
Specifications
Autoclavable Autoclavable
Color White
Holds Microtubes (1.5 mL)
Material Polypropylene
Positions 42
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Quantity 8/Cs.
Dimensions (L x W x H) 5.08 x 4.15 x 1.7 in. (12.8 x 10.5 x 4.3 cm)
Length (Metric) 12.8 cm
Length (English) 5.08 in.
Width (English) 4.15 in.
Height (English) 1.7 in.
Product Type Microcentrifuge Tube Rack
Shelves 3 Tiers
Height (Metric) 4.3 cm
Array 6 x 7
Width (Metric) 10.5 cm