Learn More
Bel-Art™ Large Pails
Product Code. 10623653 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (Metric):
7.6 L
13.2 L
Material:
Polypropylene
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material unitSize
10623653 7.6 L Polypropylene Pack of 6
17142986 13.2 L Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10623653 Supplier Bel-Art™ Supplier No. F167750000
Only null left
Add to basket
Add to basket
Rugged pails with fitted lids/covers and steel-plated bail handles
  • Translucent white pails allow for easier viewing of fluid level
  • Polypropylene is steam autoclavable (121°C), Polyethylene is not autoclavable
  • 8-qt./7.6L pail has removable steel-plated support ring and bail handle
  • 14-qt./13.2L pail has steel-plated swinging bail handle with grip, no support ring
  • Large pails meets US Specification MIL-P36257A
Specifications
Product Type Pail
Capacity (Metric) 7.6 L
Graduations None
Material Polypropylene
Autoclavable Autoclavable
Capacity (English) 8 qt.
Color White
Description 8 qt. Pail
Handle Material Plated steel
Height (English) 10 in.
Height (Metric) 25 cm
Includes Removable support ring and steel plated bail handle, snug fitting cover