Learn More
Bel-Art™ Polystop™ Dropper Bottles
Product Code. p-3206160 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
Unit Size:
Each
Product Code. 10389547
Brand: Bel-Art™ F116040030
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Prevent chipping of the glass dropper upon reinsertion into the dropper bottle.
  • The dropper easily pulls free, while the stopper remains snugly in the neck of the bottle, thus providing a safe return space for the dropper
  • Polypropylene stopper
  • Clear glass bottle has a ground T-neck, and the dropper has a soft vinyl bulb
  • Disassembles for easy cleaning
Specifications
Capacity 1 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Material Glass
Quantity 1 Ea.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Dropper Capacity 30 mL
Drop Size 0.002 oz.
Graduated No
Mouth Narrow, Threaded
Shape Cylindrical
Height (Metric) 77.5 mm
Product Type Dropping Bottle
Width (English) 1.43 in.
Height (English) 3.05 in.
Closure Material Polypropylene
Diameter (Metric) 36.3 mm
Closure Type Threaded
Autoclavable Not Autoclavable