Learn More
Bel Art™ Poxygrid™ Half-Size Test-Tube Racks
Product Code. 15500052 Shop All SP Scienceware Products
Click to view available options
Color:
Blue
Green
Orange
Positions:
20
24
36
Unit Size:
Each
Product Code. 15500052
Brand: Bel Art™ F187881603
Only null left
Add to basket
Add to basket
For use in water baths, laminar flow hoods, fume hoods or any place where space is limited
  • Epoxy coating for chemical and corrosion resistance
  • Square holes in the grid
  • Three decks with slotted grid bottom
  • Available to hold 10-13, 13-16 or 16-20 mm diameter tubes
  • Autoclavable
  • Unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand temperature range of -223°C (-369°F) to 121°C (250°F)
Specifications
Autoclavable Autoclavable
Color Orange
Holds Tubes (13 to 16 mm)
Material Epoxy-coated Steel
Positions 24
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) 13 - 16mm Tubes
Temperature (Metric) Range -23°C to +121°C
Dimensions (L x W x H) 12.5 x 9.9 x 6.4 cm (4.94 x 3.62 x 2.5 in.)
Diameter (Metric) Opening 16 mm
Product Type Test Tube Rack
Shelves 3 Tiers
Array 6 x 4