missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Swing Jaw™ Tubing Clamps

Product Code. 10478412 Shop All SP Scienceware Products
Change view
Click to view available options
Diameter (Metric) Outer Tubing:
10 mm
15 mm
20 mm
Unit Size:
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Tubing unitSize
10478412 10 mm Pack of 3
10708321 15 mm Pack of 3
10728491 20 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10478412 Supplier Bel Art™ Supplier No. H182200000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provide nonslip closures for flexible tubing

  • Made of zinc-plated steel
  • Knurled adjusting nut bears on a swinging jaw which permits full opening without removal
  • Removable from tubing without disconnecting apparatus
  • Smooth bearing surfaces will not damage tubing; tubing cannot slip out of clamp
  • Strong enough to use with heavy wall vacuum lines

Specifications

Type Tubing Clamp
Material Zinc-plated Steel
For Use With (Equipment) 10 mm (3/8 in.) O.D. flexible tubing
Diameter (English) Outer Tubing 3/8 in.
Diameter (Metric) Outer Tubing 10 mm
Color Silver
Jaw Type Swing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.