Learn More
Hanna Instruments™ Benchtop pH/mV Meter with Interval Logging
Click to view available options
Range (pH):
-2.000 to 16.000
Unit Size:
Each
Description
The HI 2215 is a microprocessor-based pH and Temperature bench meter that can also measure Oxidation Reduction Potential (ORP) in mV range with an optional electrode. A Relative mV feature is also provided. This meter is ideal for use with food testing and quality control, as well as in pharmaceutical and research laboratories.
- Temperature compensation: manual or automatic with the HI 7662 temperature probe
- Up to a five-point pH calibration can be performed using seven standard buffers and two custom buffers
- GLP capabilities allow storage and retrieval of all data regarding pH and mV calibration
- Calibration Due alarm: Alerts that too much time elapsed since the last pH calibration
- Data Logging: Store up to 100 manual logged points and 500 interval logging points
- PC Interface: Download test results via a USB port or a RS232 cable outlet
- Replaceable Electrode: Spare electrodes are available without replacing the whole meter
- Waterproof rubberised keyboard and buttons and angled wide-screen LCD
Specifications
Specifications
| Includes | HI-1131B pH electrode, HI-7662 temperature probe, HI-76404N electrode holder, Starter pack of buffer and cleaning solutions, 12 Vdc adapter and instructions |
| Accuracy (Temperature) | ±0.4°C |
| Accuracy (mV) | ±0.2 (±399.9), ±1 (±2000) |
| Accuracy (pH) | ±0.01 |
| Outputs | USB, RS-232 |
| Calibration | Automatic |
| Range (pH) | -2.000 to 16.000 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Range (mV) | ±999.9 to ±2000 |
| Resolution (mV) | 0.1 mV ; 1 mV |
| Show More |