missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bendamustine Dimer Deschloro 2’-Allyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Bendamustine Dimer Deschloro 2’-Allyl Ester |
| Analyte or Component Names | Bendamustine Dimer Deschloro 2’-Allyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H22 Cl2 F N O3 |
| Synonym | 4-(5-((2-((4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoyl)oxy)ethyl)(2-hydroxyethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic Acid Allyl Ester |
| InChI Formula | InChI=1S/C28H22Cl2FNO3/c1-28(2)21(15-22(30)17-8-11-19(29)12-9-17)26(28)27(33)35-25(16-32)18-10-13-23(31)24(14-18)34-20-6-4-3-5-7-20/h3-15,21,25-26H,1-2H3/b22-15- |
| Solution Type | Neat |
| SMILES | CC1(C)C(\C=C(/Cl)\c2ccc(Cl)cc2)C1C(=O)OC(C#N)c3ccc(F)c(Oc4ccccc4)c3 |
| IUPAC Name | [cyano-(4-fluoro-3-phenoxyphenyl)methyl] 3-[(Z)-2-chloro-2-(4-chlorophenyl)ethenyl]-2,2-dimethylcyclopropane-1-carboxylate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?