missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bendamustine Ethyl Ester, Mikromol
Description
Bendamustine Ethyl Ester, Mikromol
Specifications
Specifications
| Chemical Name or Material | Bendamustine Ethyl Ester |
| CAS | 87475-54-5 |
| Analyte or Component Names | Bendamustine Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H25 Cl2 N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, ethyl ester, Bendamustine ethyl ester, Ethyl 5-[bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazole-2-butanoate, Bendamustine Ethyl Ester |
| InChI Formula | InChI=1S/C18H25Cl2N3O2/c1-3-25-18(24)6-4-5-17-21-15-13-14(7-8-16(15)22(17)2)23(11-9-19)12-10-20/h7-8,13H,3-6,9-12H2,1-2H3 |
| Solution Type | Neat |
| SMILES | CCOC(=O)CCCc1nc2cc(ccc2n1C)N(CCCl)CCCl |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?