missing translation for 'onlineSavingsMsg'
Learn More

Bendamustine Ethyl Ester, TRC

Product Code. 30332552
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332552 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332552 Supplier Toronto Research Chemicals Supplier No. TRCB13249525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bendamustine Ethyl Ester
CAS 87475-54-5
Molecular Formula C18 H25 Cl2 N3 O2
Quantity 25 mg
Synonym 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, ethyl ester, Bendamustine ethyl ester, Ethyl 5-[bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazole-2-butanoate, Bendamustine Ethyl Ester
InChI Formula InChI=1S/C18H25Cl2N3O2/c1-3-25-18(24)6-4-5-17-21-15-13-14(7-8-16(15)22(17)2)23(11-9-19)12-10-20/h7-8,13H,3-6,9-12H2,1-2H3
SMILES CCOC(=O)CCCc1nc2cc(ccc2n1C)N(CCCl)CCCl
IUPAC Name ethyl 4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoate
Molecular Weight (g/mol) 386.32
Formula Weight 385.1324
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.