missing translation for 'onlineSavingsMsg'
Learn More

Bendamustine hydrochloride, MedChemExpress

Product Code. 16402006
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16402006

Brand: Medchem Express HYB0077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bendamustine hydrochloride (SDX-105), a purine analogue, is a DNA cross-linking agent. Bendamustine hydrochloride activats DNA-damage stress response and apoptosis. Bendamustine hydrochloride has potent alkylating, anticancer and antimetabolite properties.

Bendamustine hydrochloride (SDX-105), a purine analogue, is a DNA cross-linking agent. Bendamustine hydrochloride activats DNA-damage stress response and apoptosis. Bendamustine hydrochloride has potent alkylating, anticancer and antimetabolite properties.

Specifications

Chemical Name or Material Bendamustine hydrochloride
CAS 3543-75-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H22Cl3N3O2
Quantity 500 mg
Synonym SDX-105
Solubility Information DMSO : 100 mg/mL (253.34 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (31.67 mM; Need ultrasonic)
SMILES CN1C(C=CC(N(CCCl)CCCl)=C2)=C2N=C1CCCC(O)=O.Cl
Molecular Weight (g/mol) 394.72
Formula Weight 394.72
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.