missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benoxaprofen Methyl Ester, Mikromol™
Description
Benoxaprofen Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Benoxaprofen Methyl Ester |
| CAS | 87498-09-7 |
| Analyte or Component Names | Benoxaprofen Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H14 Cl N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-(4-Chlorophenyl)-alpha-methyl-5-benzoxazoleacetic acid methyl ester, (+/-)-Benoxaprofen methyl ester |
| InChI Formula | InChI=1S/C17H14ClNO3/c1-10(17(20)21-2)12-5-8-15-14(9-12)19-16(22-15)11-3-6-13(18)7-4-11/h3-10H,1-2H3 |
| SMILES | COC(=O)C(C)c1ccc2oc(nc2c1)c3ccc(Cl)cc3 |
| IUPAC Name | methyl 2-[2-(4-chlorophenyl)-1,3-benzoxazol-5-yl]propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?