missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benperidol N-Oxide (cis and trans), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Benperidol N-Oxide (cis and trans)
Specifications
Specifications
| Chemical Name or Material | Benperidol N-Oxide (cis and trans) |
| CAS | 118435-01-1 |
| Analyte or Component Names | Benperidol N-Oxide (cis and trans) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C22 H24 F N3 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benperidol Mixture of Imp. D and E (EP), Benperidol N-Oxide (cis and trans), 2H-Benzimidazol-2-one, 1-[1-[4-(4-fluorophenyl)-4-oxobutyl]-1-oxido-4-piperidinyl]-1,3-dihydro-, 2H-Benzimidazol-2-one, 1-[1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidinyl]-1,3-dihydro-, N-oxide, Benperidol Mixture of Impurity D and E |
| InChI Formula | InChI=1S/C22H24FN3O3/c23-17-9-7-16(8-10-17)21(27)6-3-13-26(29)14-11-18(12-15-26)25-20-5-2-1-4-19(20)24-22(25)28/h1-2,4-5,7-10,18H,3,6,11-15H2,(H,24,28) |
| SMILES | [O-][N+]1(CCCC(=O)c2ccc(F)cc2)CCC(CC1)N3C(=O)Nc4ccccc34 |
| IUPAC Name | 3-[1-[4-(4-fluorophenyl)-4-oxobutyl]-1-oxidopiperidin-1-ium-4-yl]-1H-benzimidazol-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction