Learn More
Lenz Laborglasinstrumente™ Bent Receiver Adapter with Glass Olive
Allows for the connection of a condenser to a receiving flask. Lenz Laborglasinstrumente™ Bent Receiver Adapters with Glass Olive can be directly heated and withstand typical laboratory thermal variations.
Brand:  Lenz Laborglasinstrumente™ 05275004
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11876194
  • € 54.75 / Each
Estimated Shipment: 16-04-2024
to see stock.
Description
Description
  • NS socket and NS cone
  • 105° angle
  • Vacuum connection
  • Glass olive with drain tip
  • Meets DIN 12594 standards
Specifications
Specifications
Bent Receiver Adapter with Glass Olive
Duran Glass
DIN 12594
NS 29/32