Learn More
Benzaldehyde azine, 99%, Thermo Scientific™
Product Code. 11462348
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462348 500 g 500g
11447707 25 g 25g
11452348 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11462348 Supplier Thermo Scientific Alfa Aesar Supplier No. A15721.36
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 588-68-1
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.26
MDL Number MFCD00016876
InChI Key CWLGEPSKQDNHIO-JOBJLJCHSA-N
Synonym 1,2-dibenzylidenehydrazine, n-z-benzylideneamino-1-phenylmethanimine, 1,2-di e-benzylidene hydrazine, e-n-e-benzylideneamino-1-phenylmethanimine
PubChem CID 5371793
SMILES C(=N/N=C/C1=CC=CC=C1)\C1=CC=CC=C1
Specifications
Melting Point 91°C to 95°C
Quantity 500 g
Beilstein 2208840
IUPAC Name (Z)-N-[(Z)-benzylideneamino]-1-phenylmethanimine
Formula Weight 208.27
Percent Purity 99%
Chemical Name or Material Benzaldehyde azine