missing translation for 'onlineSavingsMsg'
Learn More

Benzene 30 μg/mL in N,N-Dimethylacetamide, Dr. Ehrenstorfer

Product Code. 30319274
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319274 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319274 Supplier Dr. Ehrenstorfer Supplier No. DREA10535000DA30
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzene
CAS 71-43-2
Analyte or Component Names Benzene
Shipping Condition Room Temperature
Molecular Formula C6 H6
Concentration 30 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzene, 1,3,5-Cyclohexatriene, Benzol, Benzole, Coal naphtha, Cyclohexatriene, NSC 67315, Phene, Phenyl hydride, Pyrobenzol, Pyrobenzole, [6]Annulene
InChI Formula InChI=1S/C6H6/c1-2-4-6-5-3-1/h1-6H
SMILES c1ccccc1
IUPAC Name benzene
Solvent or Matrix N,N-Dimethylacetamide
Molecular Weight (g/mol) 78.11
Quantity 1 mL
Formula Weight 78.047 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.